Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]

Supplier: Bio X Cell
BE0009-1MG BE0009-25MG BE0009-5MG BE0009-50MG BE0009-100MG
103252-246EA 3920 USD
103252-246 103252-248 103252-250 103252-252 103572-416 76554-500 76555-718 76556-024 76556-682 76557-248
Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]
Antibodies
InVivoMab anti-mouse CD18, Clone: M18/2, Isotype: rat IgG2a, κ, Applications: in vivo LFA-1 neutralization

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: M18/2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR