Syrian Hamster IgG Isotype Control

Supplier: Bio X Cell
BE0087-5MG BE0087-1MG BE0087-25MG BE0087-50MG BE0087-100MG
103252-558EA 3920 USD
103252-558 103252-560 103252-562 103252-564 103572-782 76554-876 76555-870 76556-530 76556-938 76557-158
Syrian Hamster IgG Isotype Control
Antibodies
InVivoMab polyclonal Syrian hamster IgG, Clone: Polyclonal Syrian, Isotype: Syrian hamster IgG, Applications: in vivo administration, Functional assays, Immunohistochemistry (frozen), Immunohistochemistry (paraffin), Western blot, Immunofluorescence, Flow Cytometry

Recommended Dilution Buffer: IP0070

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR