Anti-CD54 (ICAM-1) Rat Monoclonal Antibody [clone: YN1/1.7.4]

Supplier: Bio X Cell
BE0020-1-100MG BE0020-1-1MG BE0020-1-50MG BE0020-1-5MG BE0020-1-25MG
103252-334EA 3920 USD
103252-334 103252-336 103252-338 103252-340 103572-438 76554-352 76554-618 76555-994 76556-718 76556-908
Anti-CD54 (ICAM-1) Rat Monoclonal Antibody [clone: YN1/1.7.4]
Antibodies
InVivoMab anti-mouse CD54 (ICAM-1), Clone: YN1/1.7.4, Isotype: rat IgG2b, κ, Applications: In vivo ICAM-1 neutralization

  • Recommended Dilution Buffer: IP0065

Type: Primary
Antigen: CD54 (ICAM-1)
Clonality: Monoclonal
Clone: YN1/1.7.4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR