Anti-CD103 Rat Monoclonal Antibody [clone: M290]

Supplier: Bio X Cell
BE0026-50MG BE0026-25MG BE0026-5MG BE0026-100MG BE0026-1MG
103252-382EA 3920 USD
103252-382 103252-384 103252-386 103252-388 103572-452 76554-404 76554-622 76555-394 76556-020 76556-734
Anti-CD103 Rat Monoclonal Antibody [clone: M290]
Antibodies
InVivoMab anti-mouse CD103, Clone: M290, Isotype: rat IgG2a, κ, Applications: In vivo CD103 neutralization, Immunofluorescence, Flow cytometry

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: M290
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR