Anti-ICOSL (CD275) Rat Monoclonal Antibody [clone: HK5.3]

Supplier: Bio X Cell
BE0028-5MG BE0028-50MG BE0028-25MG BE0028-1MG BE0028-100MG
103252-398EA 3920 USD
103252-398 103252-400 103252-402 103252-404 103572-456 76554-820 76556-382 76556-548 76556-608 76557-260
Anti-ICOSL (CD275) Rat Monoclonal Antibody [clone: HK5.3]
Antibodies
InVivoMab anti-mouse ICOSL (CD275), Clone: HK5.3, Isotype: rat IgG2a, κ, Applications: in vivo ICOSL neutralization

Type: Primary
Antigen: ICOSL (CD275)
Clonality: Monoclonal
Clone: HK5.3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR