Anti-IFNgammaR (CD119) Rat Monoclonal Antibody [clone: GR-20]

Supplier: Bio X Cell
BE0029-5MG BE0029-50MG BE0029-1MG BE0029-25MG BE0029-100MG
103252-406EA 3920 USD
103252-406 103252-408 103252-410 103252-412 103572-458 76554-568 76554-866 76554-896 76556-604 76556-904
Anti-IFNgammaR (CD119) Rat Monoclonal Antibody [clone: GR-20]
Antibodies
InVivoMab anti-mouse IFNγR (CD119), Clone: GR-20, Isotype: rat IgG2a, κ, Applications: in vivo IFNγR neutralization

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: IFNgammaR (CD119)
Clonality: Monoclonal
Clone: GR-20
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR