Anti-CTLA-4 (CD152) Armenian Hamster Monoclonal Antibody [clone: UC10-4F10-11]

Supplier: Bio X Cell
BE0032-25MG BE0032-50MG BE0032-5MG BE0032-1MG BE0032-100MG
103252-422EA 3920 USD
103252-422 103252-424 103252-426 103252-428 103572-462 76554-810 76555-774 76555-882 76556-636 76556-724
Anti-CTLA-4 (CD152) Armenian Hamster Monoclonal Antibody [clone: UC10-4F10-11]
Antibodies
InVivoMab anti-mouse CTLA-4 (CD152), Clone: UC10-4F10-11, Isotype: Armenian hamster IgG, Applications: in vivo CTLA-4 neutralization, in vitro CTLA-4 neutralization, Flow cytometry

  • Recommended Dilution Buffer: IP0070

Type: Primary
Antigen: CTLA-4 (CD152)
Clonality: Monoclonal
Clone: UC10-4F10-11
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR