Anti-IgG Goat Polyclonal Antibody

Supplier: Bio X Cell
BE0130-25MG BE0130-1MG BE0130-5MG BE0130-100MG BE0130-50MG
103252-708EA 3920 USD
103252-708 103252-710 103252-712 103252-714 103572-824 76555-724 76556-084 76556-928 76557-198 76557-244
Anti-IgG Goat Polyclonal Antibody
Antibodies
The polyclonal goat IgG is purified from goat serum. It is ideal for use as a non-reactive control IgG for goat IgG antibodies in most in vivo and in vitro applications.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR