Phenol Red Broth Base with Meat Extract, HiMedia Laboratories
Supplier: HiMedia Laboratories
M279-2.5KG M279-5KG M279-5KG M279-2.5KG
89225-016EA 459.46 USD
CA89225-018 89225-016 89225-018 CA89225-016
Phenol Red Broth Base with Meat Extract, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Phenol red broth base with meat extract is a highly nutritive basal medium which can be used to study fermentation of carbohydrates.