pH 2700 Benchtop Meter,Oakton®, Cole Parmer
Supplier: Avantor
101076-312EA 3144.35 USD
101076-312
pH 2700 Benchtop Meter,Oakton®, Cole Parmer
Electrochemistry Meters pH and Multi-Parameter Meters
Microprocessor based pH 2700 benchtop meter measures pH, mV, relative mV, and temperature.
  • Large display with bright backlight
  • Dynamic reading stability indication indicates when your measurement is stable
  • Password protection for both setup and calibration for security
  • Calibration-due alarm let you know when to recalibrate
  • Audible alarms alert you when reading fall out of range
  • Output data to either printer or PC