1,2-Dimyristoyl-sn-glycero-3-phosphocholine ≥99%, powder (14:0 PC)
Supplier: 0000018221
Synonyms:
14:0 PC, 1,2-Ditetradecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(tetradecanoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, DMPC, β-γ-Dimyristoyl-L-α-Lecithin, (R)-2,3-Bis(tetradecanoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate
850345P500MG
850345P1G
850345P200MG
850345P25MG
100123-992EA
253.85
USD
100123-992
100122-196
100126-508
100128-920
1,2-Dimyristoyl-sn-glycero-3-phosphocholine ≥99%, powder (14:0 PC)
1,2-Dimyristoyl-sn-glycero-3-phosphocholine
- Purified by HPLC
- Protected from oxidization and hydrolysis
Formula:
C₃₆H₇₂NO₈P MW: 677.94 g/mol |
MDL Number:
MFCD00036901 CAS Number: 18194-24-6 |
Specification Test Results
Purity (HPLC) |
|
Stability | 2 years |
Storage | ‒ 20 °C |
Transition Temperature | 24 °C |
CMC | 6 nM |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...