Dimethyl sulfoxide-[D₆] (99.96% D) for NMR spectroscopy

Supplier: CAMBRIDGE ISOTOPE LABS INC MS

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulfoxide, Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulfoxide-[²H₆], Dimethyl sulphoxide-d6, Dimethyl sulfoxide-d6

DLM-34-10X0.75 DLM-34-5X0.5 DLM-34-10X0.5 DLM-34-5
101322-270EA 315 USD
101322-270 101322-272 101322-274 101322-266 101322-268 101322-262
Dimethyl sulfoxide-[D₆] (99.96% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
MDL Number: MFCD00002090
CAS Number: 2206-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR