Anti-C3 Goat Polyclonal Antibody (Fluorescein)

Supplier: MP Biomedicals
0855191
75983-036EA 542.65 USD
75983-036
Anti-C3 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Fluorescein-Conjugated Goat F(ab')2 Fragment to Human Complement C3 is the lyophilized powder of fluorescein-5-isothiocyanate (FITC "Isomer I")-conjugated goat F(ab')2 fragment to human complement C3 and buffer salts.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacteria infection.

The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized by equivalence-point precipitation. The F(ab')2 fragments are tested for purity and specificity at a minimum of 20 mg/mL using immunoelectrophoresis.

Tissue staining, blot immunostaining

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR