Anti-SNCA Mouse Monoclonal Antibody [clone: LB509]

Supplier: Enzo Life Sciences
ABS218-0500 ABS218-0200
76002-804EA 1738.55 USD
76002-804 76002-806
Anti-SNCA Mouse Monoclonal Antibody [clone: LB509]
Antibodies
Recommended Applications: ELISA, IHC, WB

Species reactivity: Human

Type: Primary
Antigen: SNCA
Clonality: Monoclonal
Clone: LB509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Xenopus, Mammalian, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR