Mouse Recombinant Meteorin-like (from HEK293 cells)
Supplier: 0000027161
AG-40B-0149-C010 AG-40B-0149-3010
102980-314EA 263.08 USD
102980-314 102980-316
Mouse Recombinant Meteorin-like (from HEK293 cells)
Proteins and Peptides
Meteorin-like (cometin; subfatin) is a potent new neurotrophic factor promoting neurite outgrowth from dorsal root ganglion cells. Meteorin-like is also a novel adipokine expressed by adipose tissue being downregulated upon caloric restriction. Meteorin-like is secreted by muscles during exercise and converts white adipose cells into brown fat tissue.