GDC-0941 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulphonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine, 4-[2-(1H-Indazol-4-yl)-6-[(4-methylsulphonylpiperazin-1-yl)methyl]thieno[3,2-d]pyrimidin-4-yl]morpholine, Pictilisib, 4-[2-(1H-Indazol-4-yl)-6-[(4-methylsulfonylpiperazin-1-yl)methyl]thieno[3,2-d]pyrimidin-4-yl]morpholine, 4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulfonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine

CHM130-0010 CHM130-0050
76003-024EA 160.23 USD
76003-024 76003-026
GDC-0941 ≥99% (by HPLC)
GDC-0941

Cell-permeable, potent inhibitor of PI3 kinase IA and IB


A cell-permeable, potent, ATP-competitive inhibitor against class IA and IB PI3 kinases. GDC-0941 inhibits the growth of certain types of cancer cells.


Antiproliferative.

Formula: C₂₃H₂₇N₇O₃S₂
MDL Number: MFCD11616196
CAS Number: 957054-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf2a43debc7b3a206d5bf993b6b8b72e5118017&vid=eb03a044a24c48a3cedd9c95c710ec0e50cf6016&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR