Pikromycin (from Streptomyces species) ≥95% (by HPLC)
Supplier: 0000027161
BVT-0400-M001
BVT-0400-M005
102988-966EA
110.77
USD
102988-966
102988-968
Pikromycin (from Streptomyces species) ≥95% (by HPLC)
Pikromycin
Pikromycin is a 14-membered, macrolide antibiotic.
- PREP inhibitor
- Active against gram-positive bacteria
Active against Gram-positive bacteria including mycobacteria. Prolyl endopeptidase (PREP) inhibitor.
White crystalline solid
Soluble in DMSO
Storage Temperature:
Freezer |
MDL Number:
MFCD00135618 CAS Number: 19721-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...