Super Broth, Teknova
Supplier: TEKNOVA INC
S0140 S0145
100217-180EA 223.9 USD
100217-180 100218-072 100219-646 100219-876
Super Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For use in propagation bacteriophage lambda.
  • Use 57 grams per liter to reconstitute
  • Ambient shipping conditions
  • Store at room temperature
Includes 3.2% tryptone, 2.0% yeast extract, and 0.5% sodium chloride, 0.99% Pottassium Phosphate, Dibasic.