LB Media Agar, Teknova
Supplier: TEKNOVA INC
L9110 L9115 L9105
100218-380EA 2194.43 USD
100218-380 100217-750 100217-986 100216-806
LB Media Agar, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of E
  • Use 40 grams per Liter to reconstitute
  • Ambient shipping conditions
  • Store at room temperature
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), 1.0% NaCl (10.0 g/L), and 1.5% Agar (15.0 g/L).
coli and other enteric bacteria.