GoatThroat Food Grade Pumps, Westcott Distribution
Supplier: Westcott Distribution
FEP.7600 GT100 GT200 GT200S GT300
103348-872EA 1126.71 USD
103348-872 103348-902 103348-904 103348-906 103348-908
GoatThroat Food Grade Pumps, Westcott Distribution
Pumps Drum/Transfer Pumps
This maintenance-free, environmentally sound, chemical drum pump allows fast, safe transfer of more than 1400 Chemicals.
  • Works as a hand pump or pneumatically with compressors/shop air
  • Uses low pressure to deliver fluids (<5psi)
  • Delivers fluid in a steady stream with no pulsing or fluid shear
  • Has finger tip control of fluid flow for accurate dispensing fluids when needed
  • Isolates the drum contents from the outside air
  • Eliminates evaporation, contamination, and VOC emissions
  • RCRA empty barrels and containers
  • Limited 1 year manufacturer's warranty
  • Siphon tube length: 40"
These engineering controls for fluids are designed to minimize the hazards associated when transferring liquids, including leaks, spills drips, and vapor emissions
The FEP and Kalrez pump is for use with chemicals which will not work with any of the standard elastomers.
GoatThroat® Pumps work by pressurizing a container to dispense liquids with one touch, as easily as using a faucet. A few strokes of the piston can pressurize any container from 2-gallon jugs to 55-gallon drums.
Life expectancy on each pump is approximately 10 years. The set up of these pumps allows low pressure (<6 PSI) to be built up in the vessel; the power of the pressure moves the fluid. Therefore, there are fewer worker injuries than with other hand pumps which require continuous, repetitive action to move fluids.
Out of the box, it fits any barrel with a 2-inch pipe thread. There are adapters available for most 5 gallon/20 liter containers. Just pump a few strokes to pressurize, then dispense precisely controlled amounts with one touch of the spring actuated tap as easily as opening and closing a faucet. Most fluids begin to move at 2PSIG . System has automatic internal pressure relief at 8psig. Can be operated by hand or as a pneumatic. Food grade.
Certifications: Meets RoHS, ELV, WEEE, EPCRA, and CA Prop. 65, Food Grade, Made in USA. Meets U.N. safety standards for dangerous goods drums.
Ordering information: Kit includes Pump, 4 ea of 10" Siphon Tubes, 3 Connectors, 1 Foot Piece, 2" NPT Barrel Fitting, Rubber Compression Fitting, Molykote 111 lubricant, Teflon Tape, 30 Page Instruction Manual.
Confirm pump and chemical compatibility before order; warranty only applies to liquids on the approved list; consider filling out Pump Selection Guide to verify all requirements; special adapters required for buttress threads on containers; special adapters required for 5 gallon/20 liter containers.
Delivery information: Ships within 7 days ARO.