Sodium metabisulfite ≥97% ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite
Sodium metabisulfite is an antioxidant reducing agent, with antimicrobial and antifungal properties. Gastric tissue studies report that sodium metabisulfite can augment lipid peroxidation and apoptosis in a dose-dependent manner.
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.
Formula:
Na₂S₂O₅ MW: 190.11 g/mol Melting Pt: >300 °C (lit.) Density: 1000…1200 kg/m³ (bulk density) Storage Temperature: Ambient |
MDL Number:
MFCD00167602 CAS Number: 7681-57-4 EINECS: 231-673-0 |
|
Specification Test Results
Physical Appearance | White Crystalline Powder |
Heavy Metals | Pb ≤0.001%; Iron (Fe) ≤0.002% |
Insoluble Matter | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...