Ponceau S, dark powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

0219064410 0219064405
IC19064410EA 58.37 USD
IC19064410 IC0219064405
Ponceau S, dark powder
Ponceau S

Ponceau S is used as protein stain for electrophoresis.

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Melting Pt: 300 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003892
CAS Number: 6226-79-5
EINECS: 228-319-2

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: l max (water) 517 - 523, 349 - 355 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR