Chlorpromazine hydrochloride

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

0219032605 0219032625 0219032680
IC19032605EA 116.35 USD
IC19032605 IC19032625 IC19032680
Chlorpromazine hydrochloride
Chlorpromazine hydrochloride

Chlorpromazine Hydrochloride is a phenothiazine antipsychotic; a D2 and H1 antagonist. It inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase and is reported to uncouple and inhibit oxidative phosphorylation at 10-3 - 10-4 M. It is possibly a photogenotoxic agent in humans. Helps to induce hypothermia in mice in the dose range of 12.5 -100 mg/kg. Chlorpromazine demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes. Phenothiazine antipsychotic; D2 dopamine receptor antagonist, H1 histamine receptor antagonist; inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase.


Chlorpromazine hydrochloride is a substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
Melting Pt: 194…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00012654
CAS Number: 69-09-0
EINECS: 200-701-3
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96d05f889707cc689ecc9092645dca9f0e3ac63e&vid=96d05f889707cc689ecc9092645dca9f0e3ac63e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Physical Appearance White Powder
Extinction Coefficient EmM= 31.5 (0.12 M HCl) (Lit.)
Loss on Drying ≤1.0%
Solubility Soluble in water (50 mg/mL - clear, colorless to faint yellow-grey solution), methanol, ethanol, chloroform; practically insoluble in ether, benzene.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR