DAF-FM diacetate ≥98%

Supplier: 0000027161

Synonyms: DAF-FM DA, 4-Amino-5-methylamino-2',7'-difluorofluorescein diacetate

CDX-A0024-M001 CDX-A0024-M005
102989-144EA 508.15 USD
102989-144 102989-146
DAF-FM diacetate ≥98%
DAF-FM diacetate

DAF-FM diacetate is an important reagent for quantification of low concentrations of nitric oxide in solution. This compound is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. With excitation/emission maxima of 495/515 nm, DAF-FM can be detected using any instrument suitable for detecting fluorescein.

Formula: C₂₅H₁₈F₂N₂O₇
Storage Temperature: Freezer
CAS Number: 254109-22-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR