GSK1120212 ≥95%

Supplier: 0000027161
Warning

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

SYN-1170-M001 SYN-1170-M005 SYN-1170-M010 SYN-1170-M050
102992-526EA 119.08 USD
102992-526 102992-528 102992-530 102992-532
GSK1120212 ≥95%
GSK1120212

GSK1120212 (JTP-74057, trametinib) is a highly specific and potent MEK1 and MEK2 inhibitor with IC(50) values of 0.92nM and 1.8nM respectively.

Formula: C₂₆H₂₃FIN₅O₄
Storage Temperature: Freezer
MDL Number: MFCD17215075
CAS Number: 871700-17-3

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR