PIK-294 ≥95%
Supplier: 0000027161
Synonyms:
2-((4-Amino-3-(3-hydroxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)methyl)-5-methyl-3-(o-tolyl)quinazolin-4(3H)-one
SYN-1098-M001
SYN-1098-M005
SYN-1098-M010
SYN-1098-M050
102992-024EA
156.46
USD
102992-024
102992-026
102992-028
102992-030
PIK-294 ≥95%
PIK-294
PIK-294 is a PI3 Kinase inhibitor that is 20- to 60-fold more potent than the parent compound, PIK-293. It is one of the most potent p110delta-selective inhibitors that has been reported, with an IC(50) of 10nM.
Formula:
C₂₈H₂₃N₇O₂ Storage Temperature: Freezer |
MDL Number:
MFCD17019338 CAS Number: 900185-02-6 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility |
Soluble in DMSO. Slightly soluble ( |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...