GDC-0941 ≥95%

Supplier: 0000027161
Warning

Synonyms: 4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulphonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine, 4-[2-(1H-Indazol-4-yl)-6-[(4-methylsulphonylpiperazin-1-yl)methyl]thieno[3,2-d]pyrimidin-4-yl]morpholine, Pictilisib, 4-[2-(1H-Indazol-4-yl)-6-[(4-methylsulfonylpiperazin-1-yl)methyl]thieno[3,2-d]pyrimidin-4-yl]morpholine, 4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulfonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine

SYN-1041-M001 SYN-1041-M005 SYN-1041-M010 SYN-1041-M050
102991-566EA 119.08 USD
102991-566 102991-568 102991-570 102991-572
GDC-0941 ≥95%
GDC-0941

GDC-0941 is a potent class I PI3 kinase (PI3K) inhibitor. It selectively binds to PI3K isoforms in an ATP-competitive manner. It has an IC(50) of 3nM, and shows slight selectivity toward p110beta (11-fold) and p110gamma (25-fold).

Formula: C₂₃H₂₇N₇O₃S₂
Storage Temperature: Freezer
MDL Number: MFCD11616196
CAS Number: 957054-30-7

Order Now

Specification Test Results

Purity 95%
Appearance/Color Solid.
Solubility Soluble in DMSO. Slightly soluble (1mg/ml) in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR