Thiazole orange ≥98%

Supplier: 0000027161
Warning

Synonyms: 1-Methyl-4-[(3-methyl-2(3H)-benzothiazolylidene)methyl]quinolinium p-tosylate

CDX-T0013-M250 CDX-T0013-G005
102991-036EA 113.54 USD
102991-036 102991-038
Thiazole orange ≥98%
Thiazole orange

Thiazol Orange is a nucleic acid stain that has found applications in many different areas. Among others it is used for reticulocyte analysis and Plasmodium species analysis as well as labeling cancer cells. Thiazole orange-labeled DNA has been used for live cell RNA imaging. Thiazole Orange has also been used for a variety of assays, PCR and flow cytometry.

Formula: C₂₆H₂₄N₂O₃S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00192499
CAS Number: 107091-89-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Orange crystalline powder.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR