7-[(2-Oxocyclopentyl)oxy]-2H-1-benzopyran-2-one ≥98%

Supplier: 0000027161
Warning

Synonyms: 7-[(2-Oxocyclopentyl)oxy]-2H-chromen-2-one

CDX-O0047-M010 CDX-O0047-M100
102990-702EA 139.85 USD
102990-702 102990-704
7-[(2-Oxocyclopentyl)oxy]-2H-1-benzopyran-2-one ≥98%
7-[(2-Oxocyclopentyl)oxy]-2H-1-benzopyran-2-one

Fluorogenic substrates for detecting Baeyer-Villiger monooxygenase.

Formula: C₁₄H₁₂O₄
Storage Temperature: Freezer
CAS Number: 877774-65-7

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR