7-(2-Oxiranylethoxy)-2H-1-benzopyran-2-one ≥98%

Supplier: 0000027161
Warning

Synonyms: 7-(2-(Oxiran-2-yl)ethoxy)-2H-chromen-2-one

CDX-O0046-M010 CDX-O0046-M100
102990-698EA 139.85 USD
102990-698 102990-700
7-(2-Oxiranylethoxy)-2H-1-benzopyran-2-one ≥98%
7-(2-Oxiranylethoxy)-2H-1-benzopyran-2-one

Fluorogenic substrate.

Formula: C₁₃H₁₂O₄
Storage Temperature: Freezer
CAS Number: 381689-75-4

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White powder.
Solubility Soluble in ethylacetate.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR