NIR-797-isothiocyanate ≥90%
Supplier: 0000027161
Synonyms:
NIR-797-ITC
CDX-N0008-M005
CDX-N0008-M025
102990-592EA
263.08
USD
102990-592
102990-594
NIR-797-isothiocyanate ≥90%
NIR-797-isothiocyanate
The reagent, a cyanine dye, can be used as a label for the determination of proteins with NIR-fluorescent technology. The absorption-maxima of the reagent lies in a region of low interference by other biomolecules. The isothiocyanate-moiety is used to link the reagent to proteins which can sensitively be detected. Experiments to lower the detection-limits of immunoassays have been done.
Formula:
C₄₅H₅₀N₃NaO₆S₄ Storage Temperature: Freezer |
CAS Number:
152111-91-6 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Blue-green powder. |
Solubility | Soluble in ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...