1-(2-Isothiocyanatoethyl)-4-[2-(3,4-dihydro-2H-1-benzopyranyl-6-yl)-5-oxazolyl]pyridinium bromide ≥95%
Supplier: 0000027161
|
Warning
|
CDX-I0035-M005
CDX-I0035-M050
CDX-I0035-M250
102990-198EA
78.92
USD
102990-198
102990-200
102990-202
1-(2-Isothiocyanatoethyl)-4-[2-(3,4-dihydro-2H-1-benzopyranyl-6-yl)-5-oxazolyl]pyridinium bromide ≥95%
1-(2-Isothiocyanatoethyl)-4-[2-(3,4-dihydro-2H-1-benzopyranyl-6-yl)-5-oxazolyl]pyridinium bromide
Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Epsilon∼
Formula:
C₂₀H₁₈BrN₃O₂S Storage Temperature: Freezer |
CAS Number:
155863-02-8 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...