1-(2-Isothiocyanatoethyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥90%

Supplier: 0000027161
Danger

Synonyms: Stain 3-I

CDX-I0032-M025 CDX-I0032-M050 CDX-I0032-M250
102990-180EA 110.77 USD
102990-180 102990-182 102990-184
1-(2-Isothiocyanatoethyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide ≥90%
1-(2-Isothiocyanatoethyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Epsilon∼ 20,000 in EtOH).

Formula: C₁₈H₁₆BrN₃O₂S
Storage Temperature: Freezer
CAS Number: 155862-91-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR