FOAA ≥90%

Supplier: 0000027161
Warning

Synonyms: Fluorescein-O-acetate

CDX-F0022-M005 CDX-F0022-M025 CDX-F0022-M250
102989-936EA 121.85 USD
102989-936 102989-938 102989-940
FOAA ≥90%
FOAA

Fluorescent label which reacts with amines after activation to give isomerically homogeneous derivatives.

Formula: C₂₂H₁₄O₇
Storage Temperature: Freezer
CAS Number: 233759-98-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPCE)
Appearance/Color Light yellow powder.
Solubility Soluble in DMSO or acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR