DSP-2 ≥98%

Supplier: 0000027161
Warning

Synonyms: 3'-Hydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-6'-yl 2-fluoro-5-nitrobenzoate

CDX-D0392-M001 CDX-D0392-M005 CDX-D0392-M010
102989-814EA 157.85 USD
102989-814 102989-816 102989-818
DSP-2 ≥98%
DSP-2

Control for H2S2 and hydrogen polysulfides probes.

Formula: C₂₇H₁₄FNO₈
Storage Temperature: Freezer
CAS Number: 1609395-31-4

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (NMR)
Appearance/Color White to yellow crystalline.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR