Diclofop ≥98%

Supplier: 0000027161
Warning

Synonyms: 2-(4-(2,4-Dichlorophenoxy)phenoxy)propionic acid

CDX-D0314-M050 CDX-D0314-M250
102989-758EA 193.85 USD
102989-758 102989-760
Diclofop ≥98%
Diclofop

Herbicide. Peroxisome proliferator in rodents with carcinogenic activity. Compound can be used as analytical reference material.

Formula: C₁₅H₁₂Cl₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00152216
CAS Number: 40843-25-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR