6,8-Difluoro-7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylic acid ≥97%

Supplier: 0000027161
Warning

Synonyms: Pacific Blue

CDX-D0079-M010 CDX-D0079-M050
102987-760EA 70.62 USD
102987-760 102987-762
6,8-Difluoro-7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylic acid ≥97%
6,8-Difluoro-7-hydroxy-2-oxo-2H-1-benzopyran-3-carboxylic acid

Fluorinated fluorescent dye. Increased resistance to photobleaching and higher quantum yields makes it a superior fluorescent dye. Pacific Blue dyes yield conjugates that are strongly fluorescent, even at neutral pH. Spectral data: Extinction 403nm/Emmission 455nm. The amine-reactive succinimidyl ester derivative can be used to create bright blue-fluorescent bioconjugates with excitation/emission maxima ~410/455 nm that are excitable by the 405 nm spectral line of the blue diode (violet) laser.

Formula: C₁₀H₄F₂O₅
Storage Temperature: Freezer
CAS Number: 215868-31-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity 97% (HPLC)
Appearance/Color Yellow to brown powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR