Benthiavalicarb isopropyl ≥99%

Supplier: 0000027161

Synonyms: 1-Methylethyl [(1S)-1-[[[(1R)-1-(6-fluoro-2-benzothiazolyl)ethyl]amino]carbonyl]-2-methylpropyl]carbamateCBNumber:CB21120099

CDX-B0243-M025 CDX-B0243-M250
102989-426EA 211.85 USD
102989-426 102989-428
Benthiavalicarb isopropyl ≥99%
Benthiavalicarb isopropyl

Reference standard.

Formula: C₁₈H₂₄FN₃O₃S
Storage Temperature: Refrigerator
CAS Number: 177406-68-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR