5(6)-Aminofluorescein (Mixture of isomers) ≥95%

Supplier: 0000027161

Synonyms: Fluoresceinamine, 5(or 6)-Amino-3′,6′-dihydroxyspiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one, 5(6)-Aminofluorescein

CDX-A0039-G002 CDX-A0039-G010
102989-156EA 210.46 USD
102989-156 102989-158
5(6)-Aminofluorescein (Mixture of isomers) ≥95%
5(6)-Aminofluorescein (Mixture of isomers)

Two isomers are produced by the usual synthesis. Most publications imply they are interchangeable when used for the preparation of the isothiocyanate for fluorescent labeling.

Formula: C₂₀H₁₃NO₅
Storage Temperature: Freezer
MDL Number: MFCD00133331
CAS Number: 27599-63-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetone or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR