Butylcycloheptylprodigiosin ≥93%

Supplier: 0000027161

Synonyms: Butyl-ortho-cycloheptylprodiginine

BVT-0423-C250 BVT-0423-M001
102988-986EA 117.69 USD
102988-986 102988-988
Butylcycloheptylprodigiosin ≥93%
Butylcycloheptylprodigiosin

Tripyrrolic pigment like prodigiosin. Antimalarial agent. Anticancer agent with multiple modes of action. Immunosuppressant in non-toxic concentrations. Bone resorption inhibitor. Antiulcer agent.

Formula: C₂₅H₃₃N₃O
Storage Temperature: Freezer
CAS Number: 352304-41-7

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces parvulus.
Purity 93% (NMR)
Appearance/Color Red solid.
Solubility Soluble in DMSO, methanol, acetone or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR