Undecylprodigiosin hydrochloride ≥95%

Supplier: 0000027161

Synonyms: Undecylprodiginine

BVT-0422-C250 BVT-0422-M001
102988-982EA 117.69 USD
102988-982 102988-984
Undecylprodigiosin hydrochloride ≥95%
Undecylprodigiosin hydrochloride

Tripyrrolic pigment like prodigiosin. Antimalarial agent. Apoptosis inducer. Anticancer agent with multiple modes of action. Immunosuppressant in non-toxic concentrations. Bone resorption inhibitor. Antiulcer agent.

Formula: C₂₅H₃₅N₃O·HCl
Storage Temperature: Freezer
CAS Number: 56247-02-0

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces parvulus.
Purity 95% (HPLC, NMR)
Appearance/Color Red solid.
Solubility Soluble in DMSO, methanol, acetone or chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR