Cyclosporin D ≥97%

Supplier: 0000027161
Danger

Synonyms: Val2-Cyclosporine

AG-CN2-0445-M001 AG-CN2-0445-M005
102988-288EA 110.77 USD
102988-288 102988-290
Cyclosporin D ≥97%
Cyclosporin D

Potent inhibitor of tumor promoting phorbol ester TPA/PMA (AG-CN2-0010) effects in vivo. Antifungal. Moderate immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A (AG-CN2-0079). Moderate inhibitor of formyl peptide-induced superoxide formation.

Formula: C₆₃H₁₁₃N₁₁O₁₂
Storage Temperature: Freezer
MDL Number: MFCD11111526
CAS Number: 63775-96-2

Order Now

Specification Test Results

Source/Host Isolated from Fusarium solani.
Purity 97% (HPLC)
Appearance/Color White solid.
Solubility Soluble in 100% ethanol, acetone or ethyl acetate; insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR