10,11-Dehydrocurvularin ≥95%
Supplier: 0000027161
AG-CN2-0165-M001
AG-CN2-0165-M005
102988-118EA
138.46
USD
102988-118
102988-120
10,11-Dehydrocurvularin ≥95%
10,11-Dehydrocurvularin
Antibiotic
Source/Host: Isolated from Aspergillus sp.
Purity: >95% (HPLC)
Appearance/Color: Off-white solid.
Solubility: Soluble in ethanol, methanol or DMSO.
Antifungal. Nematicidal. Phytotoxic. TGF-beta signaling inhibitor Anticancer compound. Strong activator of the heat shock response. Inhibitor of the appressorium formation of M. grisea
Storage Temperature:
Freezer |
CAS Number:
1095588-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...