Cycloaspeptide A ≥95%
Supplier: 0000027161
|
Warning
|
AG-CN2-0148-M001
AG-CN2-0148-M005
102988-078EA
186.92
USD
102988-078
102988-080
Cycloaspeptide A ≥95%
Cycloaspeptide A
Antibiotic
Source/Host: Isolated from Gliocladium sp. FKI-2346.
Purity: >95% (HPLC)
Appearance/Color: White crystalline.
Solubility: Soluble in DMSO, ethanol or methanol.
Antimalaria agent.
Formula:
C₃₆H₄₃N₅O₆ Storage Temperature: Freezer |
CAS Number:
109171-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...