Altenusin ≥97%

Supplier: 0000027161
Warning

AG-CN2-0143-M001 AG-CN2-0143-M005
102988-058EA 193.85 USD
102988-058 102988-060
Altenusin ≥97%
Altenusin

Antibiotic


Source/Host: Isolated from Penicillium sp.
Purity: >97% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in methanol, ethanol or DMSO.


Antifungal penicillide. Non-competitive, specific neutral sphingomyelinase (N-SMase) inhibitor. Strong pp60c-Src inhibitor. Inhibits cFMS receptor tyrosine kinase (CSF-1/m-CSF receptor tyrosine kinase) which is implicated in cancer and bone diseases. Myosin light chain kinase inhibitor. Exhibits anti-HIV-1 integrase activity. Cytotoxic activity against mouse lymphoma cells. Anti-protozoan, trypanothione reductase (TR) inhibitor. Phytotoxin.

Storage Temperature: Freezer
CAS Number: 31186-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR