Calpinactam ≥95%
Supplier: 0000027161
|
Warning
|
Synonyms:
FKI-4905
AG-CN2-0131-C250
AG-CN2-0131-M001
102988-018EA
138.46
USD
102988-018
102988-020
Calpinactam ≥95%
Calpinactam
Specific anti-mycobacterial. Inhibits growth of Mycobacteria smegmatis but also Mycobacteria tuberculosis. M. smegmatis is used as a model for mycobacterial species. It is useful for the research analysis of other Mycobacteria species in laboratory experiments. M. smegmatis is commonly used in work on the mycobacterium species, since it is a "fast grower" and non-pathogenic.
Formula:
C₃₈H₅₇N₉O₈ Storage Temperature: Freezer |
CAS Number:
1205538-83-5 |
Specification Test Results
Source/Host | Synthetic. Originally isolated from Mortierella alpina. |
Purity |
|
Appearance/Color | Off-white solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...