Bikaverin ≥95%
Supplier: 0000027161
|
Warning
|
AG-CN2-0130-C250
AG-CN2-0130-M001
102988-014EA
90
USD
102988-014
102988-016
Bikaverin ≥95%
Bikaverin
Antibiotic
Source/Host: Isolated from Fusarium sp.
Purity: >95% (TLC)
Appearance/Color: Dark red solid.
Solubility: Soluble in DMSO.
Antiprotozoal. Antifungal. Anticancer. ATP synthesis inhibitior. Haemolytic agent. Antioomycete. Spermidine-induced autoactivation inhibitor. Plasma hyaluronan-binding protein (PHBP) inhibitor (active form). Review.
Storage Temperature:
Freezer |
CAS Number:
33390-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...