11-O-Methylpseurotin A ≥95%

Supplier: 0000027161
Warning

AG-CN2-0121-C250 AG-CN2-0121-M001
102987-978EA 69.23 USD
102987-978 102987-980
11-O-Methylpseurotin A ≥95%
11-O-Methylpseurotin A

Antibacterial. Selectively inhibits a Hof1 deletion strain.

Formula: C₂₃H₂₇NO₈
Storage Temperature: Freezer
CAS Number: 956904-34-0

Order Now

Specification Test Results

Source/Host Isolated from Sporothrix sp.
Purity 95% (HPLC)
Appearance/Color White solid.
Solubility Soluble in DMSO, ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR