2-Morpholino-8-phenylchromone

Supplier: 0000027161

Synonyms: LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran

SYN-1108-M001 SYN-1108-M005 SYN-1108-M010 SYN-1108-M050
102992-090EA 74.77 USD
102992-090 102992-092 102992-094 102992-096
2-Morpholino-8-phenylchromone
2-Morpholino-8-phenylchromone

LY294002 is a selective and reversible PI3K inhibitor with an IC(50) range of 0.5 - 1µM.

Formula: C₁₉H₁₇NO₃
MDL Number: MFCD00270881
CAS Number: 154447-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR