2,2-Dimethylcyclopentanone

Supplier: 0000027161

Synonyms: 2,2-Dimethylcyclopentan-1-one

CDX-D0333-G001 CDX-D0333-G005 CDX-D0333-G050
102989-786EA 80.31 USD
102989-786 102989-788 102989-790
2,2-Dimethylcyclopentanone
2,2-Dimethylcyclopentanone

Building block for synthesis.

Formula: C₇H₁₂O
MDL Number: MFCD00049164
CAS Number: 4541-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR